Anti-KDEL Polyclonal Antibody (ATTO 390)

Toimittaja: Abnova
ABNOPAB28965EA 655 EUR
ABNOPAB28965
Anti-KDEL Polyclonal Antibody (ATTO 390)
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type:
Antigen: KDEL
Clonality: Polyclonal
Clone:
Conjugation: ATTO 390
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR