Anti-SOD2 Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))

Toimittaja: Abnova
ABNOPAB28997EA 545 EUR
ABNOPAB28997
Anti-SOD2 Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Vasta-aineet
Rabbit polyclonal antibody raised against Recombinant of Sod2.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type:
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR