Anti-IGHM Mouse Monoclonal Antibody [clone: SPM557]
Toimittaja: Abnova
ABNOMAB12094EA 440 EUR
ABNOMAB12094
Anti-IGHM Mouse Monoclonal Antibody [clone: SPM557]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant IGHM.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: SPM557
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now