Anti-IGKC Mouse Monoclonal Antibody [clone: SPM558]

Toimittaja: Abnova
ABNOMAB12095EA 440 EUR
ABNOMAB12095
Anti-IGKC Mouse Monoclonal Antibody [clone: SPM558]
Vasta-aineet
Mouse monoclonal antibody raised against native IGKC.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 1:250-1:500

Type: Primary
Antigen: IGKC
Clonality: Monoclonal
Clone: SPM558
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR