Anti-CK19 Mouse Monoclonal Antibody [clone: SPM266]

Toimittaja: Abnova
ABNOMAB12104EA 440 EUR
ABNOMAB12104
Anti-CK19 Mouse Monoclonal Antibody [clone: SPM266]
Vasta-aineet
Mouse monoclonal antibody raised against KRT19.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CK19
Clonality: Monoclonal
Clone: SPM266
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR