Anti-CK19 Mouse Monoclonal Antibody [clone: SPM561]
Toimittaja: Abnova
ABNOMAB12105EA 440 EUR
ABNOMAB12105
Anti-CK19 Mouse Monoclonal Antibody [clone: SPM561]
Vasta-aineet
Mouse monoclonal antibody raised against KRT19.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CK19
Clonality: Monoclonal
Clone: SPM561
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now