Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM528]

Toimittaja: Abnova
ABNOMAB12107EA 440 EUR
ABNOMAB12107
Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM528]
Vasta-aineet
Mouse monoclonal antibody raised against EPCAM.

Recommended Dilutions: Immunohistochemistry: 1:100-1:200; Western Blot: 1:100-1:200

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: SPM528
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR