Anti-RAB5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Toimittaja: Abnova
ABNOPAB29010EA 492 EUR
ABNOPAB29010
Anti-RAB5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type:
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR