Anti-CNTNAP1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S65-35]
Toimittaja: Abnova
ABNOMAB11899EA 545 EUR
ABNOMAB11899
Anti-CNTNAP1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S65-35]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant Cntnap1.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: CNTNAP1
Clonality: Monoclonal
Clone: S65-35
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now