Anti-SLC12A1 Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Toimittaja: Abnova
ABNOPAB29051EA 772 EUR
ABNOPAB29051
Anti-SLC12A1 Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
Recommended Dilutions: Western Blot: 1:1000
Type:
Antigen: SLC12A1
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now