Anti-CNTN1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S73-20]

Toimittaja: Abnova
ABNOMAB11954EA 545 EUR
ABNOMAB11954
Anti-CNTN1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S73-20]
Vasta-aineet
Mouse monoclonal antibody raised aginst synthetic peptide of Cntn1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: CNTN1
Clonality: Monoclonal
Clone: S73-20
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR