Anti-SCN1A Mouse Monoclonal Antibody [clone: S74-71]

Toimittaja: Abnova
ABNOMAB11962EA 440 EUR
ABNOMAB11962
Anti-SCN1A Mouse Monoclonal Antibody [clone: S74-71]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant Scn1a.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: SCN1A
Clonality: Monoclonal
Clone: S74-71
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR