Anti-GRM5 Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29072EA 655 EUR
ABNOPAB29072
Anti-GRM5 Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against recombinant Human GRM5.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: GRM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR