Anti-ITGAM Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29076EA 655 EUR
ABNOPAB29076
Anti-ITGAM Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against recombinant Mouse Itgam.

Recommended Dilutions: Immunocytochemistry Immunohistochemistry

Type: Primary
Antigen: ITGAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR