Anti-MBP Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29078EA 655 EUR
ABNOPAB29078
Anti-MBP Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against recombinant Human MBP.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR