Anti-NTN1 Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29080EA 772 EUR
ABNOPAB29080
Anti-NTN1 Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against recombinant Human NTN1.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: NTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR