Anti-IgY Goat Monoclonal Antibody (AP (Alkaline Phosphatase))
Toimittaja: Abnova
ABNOPAB29096EA 262 EUR
ABNOPAB29096
Anti-IgY Goat Monoclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Goat polyclonal antibody raised against Chicken IgY. The antibody is conjugated with Alkaline Phosphatase.
Recommended Dilutions: Western Blot: 1:1000
Type: Secondary
Antigen: IgY
Clonality: Monoclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now