Anti-IgY Goat Monoclonal Antibody (Biotin)
Toimittaja: Abnova
ABNOPAB29097EA 419 EUR
ABNOPAB29097
Anti-IgY Goat Monoclonal Antibody (Biotin)
Vasta-aineet
Goat polyclonal antibody raised against Chicken IgY. The antibody is conjugated with Biotin.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:5000
Type: Secondary
Antigen: IgY
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now