Anti-IgY Goat Monoclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abnova
ABNOPAB29099EA 223 EUR
ABNOPAB29099
Anti-IgY Goat Monoclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Goat polyclonal antibody raised against Chicken IgY. The antibody is conjugated with HRP.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:5000

Type: Secondary
Antigen: IgY
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR