Anti-IgG Chicken Monoclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abnova
ABNOPAB29101EA 262 EUR
ABNOPAB29101
Anti-IgG Chicken Monoclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Chicken polyclonal antibody raised against Goat IgG. The antibody is conjugated with HRP.

Recommended Dilutions: Western Blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR