Anti-IgG Chicken Monoclonal Antibody

Toimittaja: Abnova
ABNOPAB29102EA 165 EUR
ABNOPAB29102
Anti-IgG Chicken Monoclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against Goat IgG.

Recommended Dilutions: Immunohistochemistry: 1:250; Western Blot: 1:1000

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR