Anti-SCN8A Mouse Monoclonal Antibody (Biotin) [clone: S87A-10]

Toimittaja: Abnova
ABNOMAB11991EA 545 EUR
ABNOMAB11991
Anti-SCN8A Mouse Monoclonal Antibody (Biotin) [clone: S87A-10]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of Scn8a.

Recommended Dilutions: Immunocytochemistry (0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: SCN8A
Clonality: Monoclonal
Clone: S87A-10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR