Anti-IgY Goat Monoclonal Antibody

Toimittaja: Abnova
ABNOPAB29112EA 194 EUR
ABNOPAB29112
Anti-IgY Goat Monoclonal Antibody
Vasta-aineet
Goat polyclonal antibody raised against Chicken IgY.

Recommended Dilutions: Immunohistochemistry: 1:250; Immunoprecipitation(1:250; Western Blot: 1:250

Type: Secondary
Antigen: IgY
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR