Anti-Soy Protein Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29115EA 399 EUR
ABNOPAB29115
Anti-Soy Protein Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against soy protein.

Recommended Dilutions: Western Blot

Type: Primary
Antigen: Soy Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR