Anti-Soy Protein Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abnova
ABNOPAB29116EA 338 EUR
ABNOPAB29116
Anti-Soy Protein Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Chicken polyclonal antibody raised against soy protein.

Type:
Antigen: Soy Protein
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR