Anti-Wheat gluten Chicken Polyclonal Antibody
Toimittaja: Abnova
ABNOPAB29117EA 399 EUR
ABNOPAB29117
Anti-Wheat gluten Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against wheat gluten.
Recommended Dilutions: ELISA: 0.5-2 ?g/ml for capture; Western Blot: 0.1-0.5 ?g/ml
Type: Primary
Antigen: Wheat gluten
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now