Anti-Wheat gluten Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abnova
ABNOPAB29118EA 338 EUR
ABNOPAB29118
Anti-Wheat gluten Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Chicken polyclonal antibody raised against wheat gluten.

Recommended Dilutions: ELISA: 0.05-0.2 ?g/ml; Western Blot: 0.1-1.0 ?g/ml

Type:
Antigen: Wheat gluten
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR