Anti-GST Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abnova
ABNOPAB29119EA 338 EUR
ABNOPAB29119
Anti-GST Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Chicken polyclonal antibody raised against Glutahtione-S-Trasferase (GST).

Recommended Dilutions: ELISA: 1:7500; Western Blot: 1:500

Type:
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR