Anti-C3 Chicken Polyclonal Antibody

Toimittaja: Abnova
PAB29121
ABNOPAB29121EA 518 EUR
ABNOPAB29121
Anti-C3 Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against synthetic peptides of Bovine C3.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR