Anti-STRN4 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S88-64]
Toimittaja: Abnova
ABNOMAB12007EA 545 EUR
ABNOMAB12007
Anti-STRN4 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S88-64]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant STRN4.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: STRN4
Clonality: Monoclonal
Clone: S88-64
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now