Anti-STRN4 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S88-64]

Toimittaja: Abnova
ABNOMAB12014EA 545 EUR
ABNOMAB12014
Anti-STRN4 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S88-64]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant STRN4.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: STRN4
Clonality: Monoclonal
Clone: S88-64
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR