Anti-ACTA2 Mouse Monoclonal Antibody [clone: SPM332]
Toimittaja: Abnova
ABNOMAB12016EA 518 EUR
ABNOMAB12016
Anti-ACTA2 Mouse Monoclonal Antibody [clone: SPM332]
Vasta-aineet
Mouse monoclonal antibody raised against ACTA2.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: SPM332
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now