Anti-BAX Mouse Monoclonal Antibody [clone: SPM336]
Toimittaja: Abnova
ABNOMAB12019EA 518 EUR
ABNOMAB12019
Anti-BAX Mouse Monoclonal Antibody [clone: SPM336]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of BAX.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: SPM336
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now