Anti-BCL2 Mouse Monoclonal Antibody [clone: SPM530]

Toimittaja: Abnova
ABNOMAB12022EA 518 EUR
ABNOMAB12022
Anti-BCL2 Mouse Monoclonal Antibody [clone: SPM530]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of BCL2.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: SPM530
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR