Anti-CD68 Mouse Monoclonal Antibody [clone: SPM130]

Toimittaja: Abnova
ABNOMAB12032EA 440 EUR
ABNOMAB12032
Anti-CD68 Mouse Monoclonal Antibody [clone: SPM130]
Vasta-aineet
Mouse monoclonal antibody raised against CD68.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: SPM130
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR