Anti-CGA Mouse Monoclonal Antibody [clone: SPM552]
Toimittaja: Abnova
ABNOMAB12038EA 440 EUR
ABNOMAB12038
Anti-CGA Mouse Monoclonal Antibody [clone: SPM552]
Vasta-aineet
Mouse monoclonal antibody raised against CGA.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: SPM552
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now