Anti-CHGA Mouse Monoclonal Antibody [clone: SPM553]
Toimittaja: Abnova
ABNOMAB12041EA 440 EUR
ABNOMAB12041
Anti-CHGA Mouse Monoclonal Antibody [clone: SPM553]
Vasta-aineet
Mouse monoclonal antibody raised against CHGA.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: SPM553
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now