Anti-CHGA Mouse Monoclonal Antibody [clone: SPM339]

Toimittaja: Abnova
ABNOMAB12043EA 440 EUR
ABNOMAB12043
Anti-CHGA Mouse Monoclonal Antibody [clone: SPM339]
Vasta-aineet
Mouse monoclonal antibody raised against CHGA.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: SPM339
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR