Anti-CR1 Mouse Monoclonal Antibody [clone: SPM554]
Toimittaja: Abnova
ABNOMAB12044EA 518 EUR
ABNOMAB12044
Anti-CR1 Mouse Monoclonal Antibody [clone: SPM554]
Vasta-aineet
Mouse monoclonal antibody raised against CR1.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CR1
Clonality: Monoclonal
Clone: SPM554
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now