Anti-Clostridium difficile Binary Toxin (B Subunit) Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29150EA 772 EUR
ABNOPAB29150
Anti-Clostridium difficile Binary Toxin (B Subunit) Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against Clostridium difficile Binary Toxin (B subunit).

Type: Primary
Antigen: Clostridium difficile Binary Toxin (B Subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR