Anti-Clostridium difficile Toxin A Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29152EA 440 EUR
ABNOPAB29152
Anti-Clostridium difficile Toxin A Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against Clostridium difficile Toxin A.

Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: Clostridium difficile Toxin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR