Anti-FKBP10 Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29171EA 419 EUR
ABNOPAB29171
Anti-FKBP10 Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against native Human FKBP10.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: FKBP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR