Anti-HBA1 Chicken Polyclonal Antibody (Biotin)
Toimittaja: Abnova
ABNOPAB29177EA 338 EUR
ABNOPAB29177
Anti-HBA1 Chicken Polyclonal Antibody (Biotin)
Vasta-aineet
Chicken polyclonal antibody raised against native Human Hemoglobin with Biotin conjugation.
Type:
Antigen: HBA1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now