Anti-IgG Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: Abnova
ABNOPAB29184EA 399 EUR
ABNOPAB29184
Anti-IgG Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Chicken polyclonal antibody raised against native Human IgG with HRP conjugation.
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now