Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM534]

Toimittaja: Abnova
ABNOMAB12078EA 440 EUR
ABNOMAB12078
Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM534]
Vasta-aineet
Mouse monoclonal antibody raised against Tacstd1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: SPM534
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR