Anti-ECM1 Mouse Monoclonal Antibody [clone: SPM217]

Toimittaja: Abnova
ABNOMAB12084EA 518 EUR
ABNOMAB12084
Anti-ECM1 Mouse Monoclonal Antibody [clone: SPM217]
Vasta-aineet
Mouse monoclonal antibody raised against ECM1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: SPM217
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR