Anti-SLC5A5 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: FP5]

Toimittaja: Abnova
ABNOMAB11507EA 545 EUR
ABNOMAB11507
Anti-SLC5A5 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: FP5]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant of SLC5A5.

Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: FP5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR