Anti-PTPRC Mouse Monoclonal Antibody [clone: SPM568]
Toimittaja: Abnova
ABNOMAB12050EA 518 EUR
ABNOMAB12050
Anti-PTPRC Mouse Monoclonal Antibody [clone: SPM568]
Vasta-aineet
Mouse monoclonal antibody raised against PTPRC.
Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/106 cells; Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: SPM568
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now