Anti-PTPRC Mouse Monoclonal Antibody [clone: SPM570]
Toimittaja: Abnova
ABNOMAB12051EA 440 EUR
ABNOMAB12051
Anti-PTPRC Mouse Monoclonal Antibody [clone: SPM570]
Vasta-aineet
Mouse monoclonal antibody raised against PTPRC.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: SPM570
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now