Anti-PMEL Mouse Monoclonal Antibody [clone: SPM286]

Toimittaja: Abnova
ABNOMAB12054EA 518 EUR
ABNOMAB12054
Anti-PMEL Mouse Monoclonal Antibody [clone: SPM286]
Vasta-aineet
Mouse monoclonal antibody raised against SILV.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PMEL
Clonality: Monoclonal
Clone: SPM286
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR